missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1,3-Bis[4-(trifluoromethyl)phenyl]urea, 97%

Brand:  Alfa Aesar™ H61822.03

Code : NEW

Additional Details : CAS Number : 1960-88-9 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15418408

  • £338.35 / 1g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

1,3-Bis[4-(trifluoromethyl)phenyl]urea
348.24
2903509
1960-88-9
MFCD01908917
1-(4-Trifluoromethylphenyl)-3-(4-trifluoromethylphenyl)urea; N,N'-Di(p-trifluoromethylphenyl)urea

Specifications

1,3-Bis[4-(trifluoromethyl)phenyl]urea
Solid
348.24
2903509
Off-white
1960-88-9
97%
MFCD01908917
1-(4-Trifluoromethylphenyl)-3-(4-trifluoromethylphenyl)urea; N,N'-Di(p-trifluoromethylphenyl)urea
1g
Videos
SDS