Alfa Aesar™ 1,3-Bis[4-(trifluoromethyl)phenyl]urea, 97%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H61822.MD

UNSPSC: 12352116

Code: NEW

Additional Details:
Additional Details: CAS Number: 1960-88-9 Weight: 0.00025kg


Quantity 250mg

Product Code. 15428408

Quantity Price
1 £ 112.78 / 250mg
Estimated Shipment date
from Supplier 06-04-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 1,3-Bis[4-(trifluoromethyl)phenyl]urea
CAS 1960-88-9
Formula Weight 348.24
MDL Number MFCD01908917
Beilstein 2903509
Synonym 1-(4-Trifluoromethylphenyl)-3-(4-trifluoromethylphenyl)urea; N,N'-Di(p-trifluoromethylphenyl)urea

Specification

Formula Weight 348.24
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Physical Form Solid
MDL Number MFCD01908917
Beilstein 2903509
Chemical Name or Material 1,3-Bis[4-(trifluoromethyl)phenyl]urea
CAS 1960-88-9
Synonym 1-(4-Trifluoromethylphenyl)-3-(4-trifluoromethylphenyl)urea; N,N'-Di(p-trifluoromethylphenyl)urea
Quantity 250mg
Health Hazard 1 Warning
Assay 97%
Color Off-white
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Documentation

Technical documents

Safety and Handling

Warning

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.