missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1,3-Bis[4-(trifluoromethoxy)phenyl]urea, 97%

Brand:  Alfa Aesar™ H64164.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15461239

  • £398.56 / 5g


Estimated Shipment date
from Supplier 07-06-2018
Add to basket

Specifications

Chemical Identifiers

1,3-Bis[4-(trifluoromethoxy)phenyl]urea
364.24
MFCD00277528
1-(4-Trifluoromethoxyphenyl)-3-(4-Trifluoromethoxyphenyl)urea; N,N'-di[4-(trifluoromethoxy)phenyl]urea

Specifications

1,3-Bis[4-(trifluoromethoxy)phenyl]urea
97%
MFCD00277528
5g
Solid
364.24
1-(4-Trifluoromethoxyphenyl)-3-(4-Trifluoromethoxyphenyl)urea; N,N'-di[4-(trifluoromethoxy)phenyl]urea
Videos
SDS