missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1,3-Bis(4-fluorophenyl)urea, 97%

Brand:  Alfa Aesar™ H62461.03

Code : NEW

Additional Details : CAS Number : 370-22-9 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15438978

  • £70.28 / 1g


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

1,3-Bis(4-fluorophenyl)urea
248.23
1-(4-Fluorophenyl)-3-(4-fluorophenyl)urea
370-22-9
MFCD00017925

Specifications

1,3-Bis(4-fluorophenyl)urea
Solid
248.23
1-(4-Fluorophenyl)-3-(4-fluorophenyl)urea
1g
370-22-9
97%
MFCD00017925
190° to 192°C
Videos
SDS