missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1,3-Bis(4-fluorophenyl)urea, 97%

Brand:  Alfa Aesar™ H62461.MD

Code : NEW

Additional Details : CAS Number : 370-22-9 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15458978

  • £17.60 / 250mg


Estimated Shipment date
from Supplier 07-06-2018
Add to basket

Specifications

Chemical Identifiers

1,3-Bis(4-fluorophenyl)urea
248.23
1-(4-Fluorophenyl)-3-(4-fluorophenyl)urea
370-22-9
MFCD00017925

Specifications

1,3-Bis(4-fluorophenyl)urea
Solid
248.23
1-(4-Fluorophenyl)-3-(4-fluorophenyl)urea
250mg
370-22-9
97%
MFCD00017925
190° to 192°C
Videos
SDS