missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(2,6-diisopropylphenyl)imidazolidinium tetrafluoroborate, 97+%, Thermo Scientific™

Catalog Number p-3777537
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
282109-83-5
C27H39BF4N2
478.42
MFCD04971187
KFZBJQUHHALFSR-UHFFFAOYSA-N
1,3-bis 2,6-diisopropylphenyl-4,5-dihydroimidazolium tetrafluoroborate, 1,3-bis 2,6-di-i-propylphenyl-4,5-dihydroimidazolium tetrafluoroborate, sipr-hbf4, 1,3-bis 2,6-diisopropylphenyl-4,5-dihydro-1h-imidazol-3-ium tetrafluoroborate, 4,5-dihydro-1,3-bis 2,6-diisopropylphenyl imidazolium tetrafluoroborate, n,n'-bis 2,6-diisopropylphenyl dihydroimidazolium tetrafluoroborate, sipr.hbf4, 1,3-bis-2,6-diisopropylphenyl imidazolinium tetrafluoroborate, n,n'-2,6-diisopropylphenyl dihydroimidazolium tetrafluoroborate, 1,3-bis 2,6-di-i-propylphenyl-4,5-dihydro-imidazolium tetra-fluoroborate
11375006
1,3-bis[2,6-di(propan-2-yl)phenyl]-4,5-dihydroimidazol-1-ium;tetrafluoroborate
[B-](F)(F)(F)F.CC(C)C1=C(C(=CC=C1)C(C)C)N2CC[N+](=C2)C3=C(C=CC=C3C(C)C)C(C)C
This item is not returnable. View return policy
282109-83-5
C27H39BF4N2
478.42
MFCD04971187
KFZBJQUHHALFSR-UHFFFAOYSA-N
1,3-bis 2,6-diisopropylphenyl-4,5-dihydroimidazolium tetrafluoroborate, 1,3-bis 2,6-di-i-propylphenyl-4,5-dihydroimidazolium tetrafluoroborate, sipr-hbf4, 1,3-bis 2,6-diisopropylphenyl-4,5-dihydro-1h-imidazol-3-ium tetrafluoroborate, 4,5-dihydro-1,3-bis 2,6-diisopropylphenyl imidazolium tetrafluoroborate, n,n'-bis 2,6-diisopropylphenyl dihydroimidazolium tetrafluoroborate, sipr.hbf4, 1,3-bis-2,6-diisopropylphenyl imidazolinium tetrafluoroborate, n,n'-2,6-diisopropylphenyl dihydroimidazolium tetrafluoroborate, 1,3-bis 2,6-di-i-propylphenyl-4,5-dihydro-imidazolium tetra-fluoroborate
11375006
1,3-bis[2,6-di(propan-2-yl)phenyl]-4,5-dihydroimidazol-1-ium;tetrafluoroborate
[B-](F)(F)(F)F.CC(C)C1=C(C(=CC=C1)C(C)C)N2CC[N+](=C2)C3=C(C=CC=C3C(C)C)C(C)C

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 97.0
CAS Max % 100.0
Color White
Elemental Analysis 97.0% min.
Infrared Spectrum Authentic
Assay Percent Range 97+%
Packaging Glass bottle
Quantity 5 g
Formula Weight 478.42
Percent Purity 97+%
Physical Form Powder
Chemical Name or Material 1, 3-Bis(2, 6-diisopropylphenyl)imidazolidinium tetrafluoroborate
Show More Show Less
compliance-icons
Product Identifier
  • 1,3-Bis(2,6-diisopropylphenyl)imidazolidinium tetrafluoroborate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'