missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(2,6-diisopropylphenyl)imidazolidinium chloride, 90%, Thermo Scientific Chemicals

Catalog Number p-3777535
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
258278-25-0
C27H39ClN2
427.07
MFCD07369796
LWPXTYZKAWSRIP-UHFFFAOYSA-M
1,3-bis 2,6-diisopropylphenyl-4,5-dihydro-1h-imidazol-3-ium chloride, sipr.hcl, 1,3-bis 2,6-diisopropylphenyl-imidazolidinium-chloride, 1,3-bis 2,6-diisopropylphenyl imidazolidinium chloride, n,n'-2,6-diisopropylphenyl dihydroimidazolium chloride, 1,3-bis 2,6-diisopropylphenyl-4,5-dihydro-1??-imidazol-1-ylium chloride, 1,3-bis 2,6-diisopropylphenyl imidazolinium chloride?, 1,3-bis-2,6-diisopropylphenyl imidazolidinium chloride, 1,3-bis-2,6-diisopropylphenyl imidazolidiniumchloride, 1,3-bis 2,6-bis isopropyl phenyl imidazolin-3-ium chloride
2734918
1,3-bis[2,6-di(propan-2-yl)phenyl]-4,5-dihydroimidazol-1-ium;chloride
[Cl-].CC(C)C1=CC=CC(C(C)C)=C1N1CC[N+](=C1)C1=C(C=CC=C1C(C)C)C(C)C
This item is not returnable. View return policy
258278-25-0
C27H39ClN2
427.07
MFCD07369796
LWPXTYZKAWSRIP-UHFFFAOYSA-M
1,3-bis 2,6-diisopropylphenyl-4,5-dihydro-1h-imidazol-3-ium chloride, sipr.hcl, 1,3-bis 2,6-diisopropylphenyl-imidazolidinium-chloride, 1,3-bis 2,6-diisopropylphenyl imidazolidinium chloride, n,n'-2,6-diisopropylphenyl dihydroimidazolium chloride, 1,3-bis 2,6-diisopropylphenyl-4,5-dihydro-1??-imidazol-1-ylium chloride, 1,3-bis 2,6-diisopropylphenyl imidazolinium chloride?, 1,3-bis-2,6-diisopropylphenyl imidazolidinium chloride, 1,3-bis-2,6-diisopropylphenyl imidazolidiniumchloride, 1,3-bis 2,6-bis isopropyl phenyl imidazolin-3-ium chloride
2734918
1,3-bis[2,6-di(propan-2-yl)phenyl]-4,5-dihydroimidazol-1-ium;chloride
[Cl-].CC(C)C1=CC=CC(C(C)C)=C1N1CC[N+](=C1)C1=C(C=CC=C1C(C)C)C(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 88.0
CAS Max % 100.0
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 88% min. (Argentometry)
Packaging Glass bottle
Quantity 1 g
Formula Weight 427.07
Percent Purity 90%
Physical Form Crystalline Powder
Chemical Name or Material 1, 3-Bis(2, 6-diisopropylphenyl)imidazolidinium chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'