missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(2,4,6-trimethylphenyl)imidazolium chloride, 95%, ACROS Organics™

Brand:  Acros Organics 356190050

Code : 29

Additional Details : CAS Number : 141556-45-8 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10299743

  • £185.80 / 5g


EU Stock 3
Add to basket


Specifications

Chemical Identifiers

1,3-Bis(2,4,6-trimethylphenyl)imidazolium chloride
94.0
≥94%
C21H25ClN2
MFCD02684541
141556-45-8
100.0
95%
340.9

Specifications

1,3-Bis(2,4,6-trimethylphenyl)imidazolium chloride
94.0
Authentic
Powder
95%
340.9
Off-white to beige
141556-45-8
100.0
Glass bottle
≥94%
C21H25ClN2
MFCD02684541
5g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 1,3-Bis(2,4,6-trimethylphenyl)imidazolium chloride

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS