Learn More
1,3-Bis(2,4,6-trimethylphenyl)imidazolium chloride, 95%, ACROS Organics™
Brand: Acros Organics 356190010
Code : 29
Additional Details : CAS Number : 141556-45-8 Weight : 0.05100kg
Packaging | Glass bottle |
---|---|
Quantity | 1g |
Chemical Identifiers
141556-45-8 | |
340.9 | |
OTOSIXGMLYKKOW-UHFFFAOYSA-M | |
2734211 | |
CC1=CC(=C(C(=C1)C)N2C=C[N+](=C2)C3=C(C=C(C=C3C)C)C)C.[Cl-] |
C21H25ClN2 | |
MFCD02684541 | |
1,3-bis 2,4,6-trimethylphenyl imidazolium chloride, 1,3-dimesityl-1h-imidazol-3-ium chloride, 1,3-dimesitylimidazolium chloride, 1,3-bis mesityl imidazolium chloride, imes.hcl, 1,3-bis 2,4,6-trimethylphenyl imidazoliumchloride, 1,3-2,4,6-trimethylphenyl imidazolium chloride, 1,3-bis-2,4,6-trimethylphenyl-1h-imidazolium chloride, 1,3-bis-2,4,6-trimethyl-phenyl-3h-imidazol-1-ium chloride, 1,3-dihydro-1,3-dimesityl-2h-imidazol-2-ylidene monohydrochloride | |
1,3-bis(2,4,6-trimethylphenyl)imidazol-1-ium;chloride |
Specifications
1,3-Bis(2,4,6-trimethylphenyl)imidazolium chloride | |
Glass bottle | |
1g | |
94.0 | |
94% min. (Argentometry) | |
MFCD02684541 | |
OTOSIXGMLYKKOW-UHFFFAOYSA-M | |
1,3-bis(2,4,6-trimethylphenyl)imidazol-1-ium;chloride | |
2734211 | |
Powder |
Authentic | |
White to Beige | |
141556-45-8 | |
100.0 | |
C21H25ClN2 | |
1,3-bis 2,4,6-trimethylphenyl imidazolium chloride, 1,3-dimesityl-1h-imidazol-3-ium chloride, 1,3-dimesitylimidazolium chloride, 1,3-bis mesityl imidazolium chloride, imes.hcl, 1,3-bis 2,4,6-trimethylphenyl imidazoliumchloride, 1,3-2,4,6-trimethylphenyl imidazolium chloride, 1,3-bis-2,4,6-trimethylphenyl-1h-imidazolium chloride, 1,3-bis-2,4,6-trimethyl-phenyl-3h-imidazol-1-ium chloride, 1,3-dihydro-1,3-dimesityl-2h-imidazol-2-ylidene monohydrochloride | |
CC1=CC(=C(C(=C1)C)N2C=C[N+](=C2)C3=C(C=C(C=C3C)C)C)C.[Cl-] | |
340.9 | |
340.9 | |
95% |
Safety and Handling
- 1,3-Bis(2,4,6-trimethylphenyl)imidazolium chloride
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.