missing translation for 'onlineSavingsMsg'
Learn More

1,3-Benzothiazole-6-sulfonyl chloride, ≥95%, Maybridge

Brand:  Maybridge CC05903DA

Code : 31

Additional Details : CAS Number : 181124-40-3 Weight : 0.05100kg Transport : UN number : 3261 Chem class : 8 Pack group : III

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10384372

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

1, 3-Benzothiazole-6-sulfonyl chloride
94.5
95%
C7H4ClNO2S2
MFCD03086100
181124-40-3
100.0
95%
233.7

Specifications

1, 3-Benzothiazole-6-sulfonyl chloride
94.5
Amber Glass Bottle
95%
233.7
1g
181124-40-3
100.0
95%
C7H4ClNO2S2
MFCD03086100
Videos
SDS