missing translation for 'onlineSavingsMsg'
Learn More

1,3-Benzodioxole-4-carboxylic acid, ≥97%, Thermo Scientific™

Catalog Number p-3174729
Click to view available options
Quantity:
250 mg
This item is not returnable. View return policy
5768-39-8
C8H6O4
166.132
MFCD01076411
DBUAYOWCIUQXQW-UHFFFAOYSA-N
benzo d 1,3 dioxole-4-carboxylic acid, 2h-1,3-benzodioxole-4-carboxylic acid, 1,3-benzodioxole-4-carboxylicacid, 2,3-methylenedioxybenzoic acid, 2,3-methylenedioxy benzoic acid, o-piperonylic acid, 2-methylenedioxybenzoic acid, 4-carboxy-1,3-benzodioxole, 1-carboxy-methylenedioxybenzene, benzo 1,3 dioxole-4-carboxylic acid
304832
1,3-benzodioxole-4-carboxylic acid
C1OC2=CC=CC(=C2O1)C(=O)O
This item is not returnable. View return policy
5768-39-8
C8H6O4
166.132
MFCD01076411
DBUAYOWCIUQXQW-UHFFFAOYSA-N
benzo d 1,3 dioxole-4-carboxylic acid, 2h-1,3-benzodioxole-4-carboxylic acid, 1,3-benzodioxole-4-carboxylicacid, 2,3-methylenedioxybenzoic acid, 2,3-methylenedioxy benzoic acid, o-piperonylic acid, 2-methylenedioxybenzoic acid, 4-carboxy-1,3-benzodioxole, 1-carboxy-methylenedioxybenzene, benzo 1,3 dioxole-4-carboxylic acid
304832
1,3-benzodioxole-4-carboxylic acid
C1OC2=CC=CC(=C2O1)C(=O)O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 166.13
Percent Purity 97%
Chemical Name or Material 1, 3-Benzodioxole-4-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'