missing translation for 'onlineSavingsMsg'
Learn More

1,3-Benzodioxol-5-ylmethyl isothiocyanate, ≥95%, Maybridge

Brand:  Maybridge S03337DE

Code : 31

Additional Details : CAS Number : 4430-47-1 Weight : 0.05500kg Transport : UN number : 2810 Chem class : 1 Pack group : III


Quantity 5g

 View more versions of this product

Product Code. 10556615

  • £15.00 / 5g


Estimated Shipment date
from Supplier 26-02-2018
Add to basket


Specifications

Chemical Identifiers

1, 3-Benzodioxol-5-ylmethyl isothiocyanate
95.0
95%
193.23
4430-47-1
95%
C9H7NO2S
MFCD00041217

Specifications

1, 3-Benzodioxol-5-ylmethyl isothiocyanate
95.0
95%
193.23
1.32g/mL
4430-47-1
95%
C9H7NO2S
MFCD00041217
5g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 1,3-Benzodioxol-5-ylmethyl isothiocyanate

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS