missing translation for 'onlineSavingsMsg'
Learn More

1,3-Benzodioxol-4-ylmethanol, 97%, Thermo Scientific™

Catalog Number p-2694628
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
769-30-2
C8H8O3
152.149
MFCD02681980
XVCMMPXFVAHHQN-UHFFFAOYSA-N
benzo d 1,3 dioxol-4-ylmethanol, 1,3-benzodioxole-4-methanol, 2h-1,3-benzodioxol-4-ylmethanol, benzo 1,3 dioxol-4-yl-methanol, 1,3-benzodioxol-4-yl-methanol, 1,3-benzodioxol-4-yl methanol, 2,3-methylenedioxy benzyl alcohol, 4-hydroxymethyl-1,3-benzodioxole, benzo d 1,3 dioxol-7-yl methanol, 2h-benzo d 1,3-dioxolen-4-ylmethan-1-ol
2776187
1,3-benzodioxol-4-ylmethanol
C1OC2=CC=CC(=C2O1)CO
This item is not returnable. View return policy
769-30-2
C8H8O3
152.149
MFCD02681980
XVCMMPXFVAHHQN-UHFFFAOYSA-N
benzo d 1,3 dioxol-4-ylmethanol, 1,3-benzodioxole-4-methanol, 2h-1,3-benzodioxol-4-ylmethanol, benzo 1,3 dioxol-4-yl-methanol, 1,3-benzodioxol-4-yl-methanol, 1,3-benzodioxol-4-yl methanol, 2,3-methylenedioxy benzyl alcohol, 4-hydroxymethyl-1,3-benzodioxole, benzo d 1,3 dioxol-7-yl methanol, 2h-benzo d 1,3-dioxolen-4-ylmethan-1-ol
2776187
1,3-benzodioxol-4-ylmethanol
C1OC2=CC=CC(=C2O1)CO
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 152.15
Percent Purity 97%
Chemical Name or Material 1, 3-Benzodioxol-4-ylmethanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'