missing translation for 'onlineSavingsMsg'
Learn More

1,3-acetonedicarboxylic acid, 96%, Thermo Scientific Chemicals

Catalog Number 10733371
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic drum
This item is not returnable. View return policy
542-05-2
C5H6O5
146.1
MFCD00002711
OXTNCQMOKLOUAM-UHFFFAOYSA-N
1,3-acetonedicarboxylic acid, 3-oxoglutaric acid, acetonedicarboxylic acid, pentanedioic acid, 3-oxo, acetone-1,3-dicarboxylic acid, 3-ketoglutaric acid, unii-ih7p7wo21p, 1,3-acetonedicarboxylic acid; 3-ketoglutaric acid, ih7p7wo21p, acetondicarbonsaure
68328
3-oxopentanedioic acid
C(C(=O)CC(=O)O)C(=O)O
This item is not returnable. View return policy
542-05-2
C5H6O5
146.1
MFCD00002711
OXTNCQMOKLOUAM-UHFFFAOYSA-N
1,3-acetonedicarboxylic acid, 3-oxoglutaric acid, acetonedicarboxylic acid, pentanedioic acid, 3-oxo, acetone-1,3-dicarboxylic acid, 3-ketoglutaric acid, unii-ih7p7wo21p, 1,3-acetonedicarboxylic acid; 3-ketoglutaric acid, ih7p7wo21p, acetondicarbonsaure
68328
3-oxopentanedioic acid
C(C(=O)CC(=O)O)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Melting Point 133°C
Color White
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass Bottle
Linear Formula HO2CCH2COCH2CO2H
Quantity 100 g
Beilstein 03,789
Fieser 11,2
Merck Index 15,67
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,slightly soluble in ethyl acetate and ether,insoluble in benzene,chloroform,ligroin,decomposes in hot water,acids,alkalies to co2 an,d acetone
Formula Weight 146.1
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 1, 3-Acetonedicarboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 1,3-Acetonedicarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'