missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3,5-Tris(chloromethyl)-2,4,6-trimethylbenzene, 97%, Thermo Scientific Chemicals
30.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 mg
1 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Quantity | 250 mg |
UN Number | UN3261 |
Beilstein | 2369523 |
Synonym | 1,3,5-tris chloromethyl-2,4,6-trimethylbenzene, 1,3,5-tris-chloromethyl-2,4,6-trimethyl-benzene, benzene, 1,3,5-tris chloromethyl-2,4,6-trimethyl, ksc224a3n, tris-chloromethyl-mesitylene, 2,4,6-tris chloromethyl mesitylene, tris chloromethyl-2,4,6-trimethylbenzene, 1,3,5-trimethyl-2,4,6-tris chloromethyl benzene, benzene,3,5-tris chloromethyl-2,4,6-trimethyl, 1,3,5-tris chloromethyl-2,4,6-trimethyl-benzene |
SMILES | CC1=C(C(=C(C(=C1CCl)C)CCl)C)CCl |
PubChem CID | 291188 |
Formula Weight | 265.6 |
Percent Purity | 97% |
Physical Form | Powder |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'