missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Tris(4-bromophenyl)benzene, 97%, Thermo Scientific Chemicals

Catalog Number 15487797
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
7511-49-1
C24H15Br3
543.096
MFCD00362911
HJQRITCAXSBOPC-UHFFFAOYSA-N
1,3,5-tris 4-bromophenyl benzene, 4,4-dibromo-5'-4-bromophenyl-1,1':3',1-terphenyl, 1,1':3',1-terphenyl, 4,4-dibromo-5'-4-bromophenyl, 4'-bromo-3,5-bis 4-bromophenyl-1,1'-biphenyl, acmc-209ox3, ksc496e7d, 1,3,5-tris p-bromophenyl benzene, 1,3,5-tri 4-bromophenyl benzene, 4,4-dibromo-5'-4-bromophenyl-1,1'
232761
1,3,5-tris(4-bromophenyl)benzene
C1=CC(=CC=C1C2=CC(=CC(=C2)C3=CC=C(C=C3)Br)C4=CC=C(C=C4)Br)Br
This item is not returnable. View return policy
7511-49-1
C24H15Br3
543.096
MFCD00362911
HJQRITCAXSBOPC-UHFFFAOYSA-N
1,3,5-tris 4-bromophenyl benzene, 4,4-dibromo-5'-4-bromophenyl-1,1':3',1-terphenyl, 1,1':3',1-terphenyl, 4,4-dibromo-5'-4-bromophenyl, 4'-bromo-3,5-bis 4-bromophenyl-1,1'-biphenyl, acmc-209ox3, ksc496e7d, 1,3,5-tris p-bromophenyl benzene, 1,3,5-tri 4-bromophenyl benzene, 4,4-dibromo-5'-4-bromophenyl-1,1'
232761
1,3,5-tris(4-bromophenyl)benzene
C1=CC(=CC=C1C2=CC(=CC(=C2)C3=CC=C(C=C3)Br)C4=CC=C(C=C4)Br)Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 261°C to 265°C
Color Brown to Yellow
Quantity 1 g
Formula Weight 543.1
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Tris(4-bromophenyl)benzene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'