missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trimethyl-1H-pyrazole-4-sulfonyl chloride, 97%, Maybridge

Brand:  Maybridge CC14703DA

Code : 31

Additional Details : CAS Number : 59340-27-1 Weight : 0.05100kg Transport : UN number : 1759 Chem class : 1a Pack group : III

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10232782

  • £72.00 / 1g


Estimated Shipment date
from Supplier 26-02-2018
Add to basket

Specifications

Chemical Identifiers

1, 3, 5-Trimethyl-1H-pyrazole-4-sulfonyl chloride
96.5
97%
C6H9ClN2O2S
MFCD02180944
59340-27-1
100.0
97%
208.67

Specifications

1, 3, 5-Trimethyl-1H-pyrazole-4-sulfonyl chloride
59340-27-1
100.0
97%
C6H9ClN2O2S
MFCD02180944
1g
96.5
Amber Glass Bottle
97%
208.67
Videos
SDS