missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trimethyl-1H-pyrazole-4-carbaldehyde, Thermo Scientific™

Click to view available options
Quantity:
1g
5g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
2644-93-1
C7H10N2O
138.17
HMTUBXVXHHITGO-UHFFFAOYSA-N
1,3,5-trimethyl-1h-pyrazole-4-carbaldehyde, 1,3,5-trimethyl-1h-pyrazole-4-carboxaldehyde, 1h-pyrazole-4-carboxaldehyde, 1,3,5-trimethyl, n-methyl-3,5-dimethyl-4-formylpyrazole, 1,3,5-trimethyl-1h-pyrazole-4-carbaldehyde?, akos pao-0265, timtec-bb sbb000012, acmc-1cq3n, art-chem-bb b006543, ksc496q8h
736515
1,3,5-trimethylpyrazole-4-carbaldehyde
CC1=C(C(=NN1C)C)C=O
This item is not returnable. View return policy
2644-93-1
C7H10N2O
138.17
HMTUBXVXHHITGO-UHFFFAOYSA-N
1,3,5-trimethyl-1h-pyrazole-4-carbaldehyde, 1,3,5-trimethyl-1h-pyrazole-4-carboxaldehyde, 1h-pyrazole-4-carboxaldehyde, 1,3,5-trimethyl, n-methyl-3,5-dimethyl-4-formylpyrazole, 1,3,5-trimethyl-1h-pyrazole-4-carbaldehyde?, akos pao-0265, timtec-bb sbb000012, acmc-1cq3n, art-chem-bb b006543, ksc496q8h
736515
1,3,5-trimethylpyrazole-4-carbaldehyde
CC1=C(C(=NN1C)C)C=O
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5g
Formula Weight 138.17
Percent Purity 97%
Chemical Name or Material 1, 3, 5-Trimethyl-1H-pyrazole-4-carbaldehyde