missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Triisopropylbenzene, 95%, Thermo Scientific Chemicals

Catalog Number p-205590
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
717-74-8
C15H24
204.35
MFCD00008890
VUMCUSHVMYIRMB-UHFFFAOYSA-N
1,3,5-triisopropylbenzene, 2,4,6-triisopropylbenzene, benzene, 1,3,5-tris 1-methylethyl, 1,3,5-tris propan-2-yl benzene, unii-fr9y346wpb, benzene, 1,3,5-triisopropyl, fr9y346wpb, benzene, tris 1-methylethyl, pubchem13720, 1,5-triisopropylbenzene
12860
1,3,5-tri(propan-2-yl)benzene
CC(C)C1=CC(=CC(=C1)C(C)C)C(C)C
This item is not returnable. View return policy
717-74-8
C15H24
204.35
MFCD00008890
VUMCUSHVMYIRMB-UHFFFAOYSA-N
1,3,5-triisopropylbenzene, 2,4,6-triisopropylbenzene, benzene, 1,3,5-tris 1-methylethyl, 1,3,5-tris propan-2-yl benzene, unii-fr9y346wpb, benzene, 1,3,5-triisopropyl, fr9y346wpb, benzene, tris 1-methylethyl, pubchem13720, 1,5-triisopropylbenzene
12860
1,3,5-tri(propan-2-yl)benzene
CC(C)C1=CC(=CC(=C1)C(C)C)C(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Color Colorless
Density 0.8400g/mL
Boiling Point 232°C to 236°C
Flash Point 86°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula C6H3[CH(CH3)2]3
Refractive Index 1.4874 to 1.4894
Quantity 25 mL
Beilstein 05, 458
Specific Gravity 0.84
Formula Weight 204.35
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 1, 3, 5-Triisopropylbenzene, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'