missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1,3,5-Tri-n-propylhexahydro-1,3,5-triazine, 96%

Brand:  Alfa Aesar™ H64078.03

Code : NEW

Additional Details : CAS Number : 13036-81-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15471119

  • £38.30 / 1g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

1,3,5-Tri-n-propylhexahydro-1,3,5-triazine
213.37
111423
13036-81-2
MFCD00040461
Hexahydro-1,3,5-tri-n-propyl-1,3,5-triazine; 1,3,5-Tri-n-propyl-1,3,5-triazinane

Specifications

1,3,5-Tri-n-propylhexahydro-1,3,5-triazine
94°C (0.7mmHg)
1g
1.4660
96%
MFCD00040461
Hexahydro-1,3,5-tri-n-propyl-1,3,5-triazine; 1,3,5-Tri-n-propyl-1,3,5-triazinane
0.880g/mL
Colorless
13036-81-2
Liquid
213.37
111423
Videos
SDS