missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(3,5-Dimethylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea, 97%

Brand:  Alfa Aesar™ H64765.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15429136

  • £351.92 / 5g


Estimated Shipment date
from Supplier 09-07-2018
Add to basket

Specifications

Chemical Identifiers

1-(3,5-Dimethylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea
324.3
MFCD06167925

Specifications

1-(3,5-Dimethylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea
Solid
324.3
5g
97%
MFCD06167925
Videos
SDS