missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(3,5-Difluorophenyl)-3-phenylurea, 95%

Brand:  Alfa Aesar™ H61985.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15448578

  • £73.56 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

1-(3,5-Difluorophenyl)-3-phenylurea
248.23
MFCD18185585

Specifications

1-(3,5-Difluorophenyl)-3-phenylurea
1g
95%
MFCD18185585
White
Solid
248.23
Videos
SDS