missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(3,5-Difluorophenyl)-3-[4-(trifluoromethyl)phenyl]urea, 97%

Brand:  Alfa Aesar™ H61697.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15416716

  • £73.56 / 1g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

1-(3,5-Difluorophenyl)-3-[4-(trifluoromethyl)phenyl]urea
316.23
MFCD26143296

Specifications

1-(3,5-Difluorophenyl)-3-[4-(trifluoromethyl)phenyl]urea
1g
97%
MFCD26143296
White to pale yellow
Solid
316.23
Videos
SDS