Learn More
Alfa Aesar™ 1-(3,5-Dichlorophenyl)-3-[4-(trifluoromethyl)phenyl]urea, 97%
Brand: Alfa Aesar™ H61214.03
Code : NEW
Additional Details : CAS Number : 23750-74-5 Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
1-(3,5-Dichlorophenyl)-3-[4-(trifluoromethyl)phenyl]urea | |
349.14 | |
3068853 |
23750-74-5 | |
MFCD07063836 |
Specifications
1-(3,5-Dichlorophenyl)-3-[4-(trifluoromethyl)phenyl]urea | |
23750-74-5 | |
97% | |
MFCD07063836 |
1g | |
Solid | |
349.14 | |
3068853 |