missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-[3,5-Bis(trifluoromethyl)phenyl]-3-[4-(trifluoromethyl)phenyl]urea, 97%

Brand:  Alfa Aesar™ H61731.03

Code : NEW

Additional Details : CAS Number : 23747-71-9 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15416746

  • £68.26 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

1-[3,5-Bis(trifluoromethyl)phenyl]-3-[4-(trifluoromethyl)phenyl]urea
416.24
3075996
23747-71-9
MFCD26143299

Specifications

1-[3,5-Bis(trifluoromethyl)phenyl]-3-[4-(trifluoromethyl)phenyl]urea
Solid
416.24
3075996
23747-71-9
97%
MFCD26143299
1g
Videos
SDS