missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-[3,5-Bis(trifluoromethyl)phenyl]-3-[4-chloro-3-(trifluoromethyl)phenyl]urea, 97%

Brand:  Alfa Aesar™ H65834.03

Code : NEW

Additional Details : CAS Number : 4528-83-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15405775

  • £132.50 / 1g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

1-[3,5-Bis(trifluoromethyl)phenyl]-3-[4-chloro-3-(trifluoromethyl)phenyl]urea
4528-83-0
450.69
MFCD27975824

Specifications

1-[3,5-Bis(trifluoromethyl)phenyl]-3-[4-chloro-3-(trifluoromethyl)phenyl]urea
Solid
450.69
1g
4528-83-0
97%
MFCD27975824
Videos
SDS