missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Benzenetricarboxylic acid triallyl ester, 98%, Thermo Scientific Chemicals

Catalog Number 10664935
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
17832-16-5
C18H18O6
330.33
MFCD00008647
VOSUIKFOFHZNED-UHFFFAOYSA-N
triallyl trimesate, triallyl benzene-1,3,5-tricarboxylate, triallyl 1,3,5-benzenetricarboxylate, triallylbenzene-1,3,5-tricarboxylate, 1,3,5-benzenetricarboxylic acid, tri-2-propenyl ester, 1,3,5-benzenetricarboxylic acid, 1,3,5-tri-2-propen-1-yl ester, 1,3,5-benzenetricarboxylic acid triallyl ester, 1,3,5-tris prop-2-en-1-yl benzene-1,3,5-tricarboxylate, triallyltrimesate, acmc-1c8hf
87328
tris(prop-2-enyl) benzene-1,3,5-tricarboxylate
C=CCOC(=O)C1=CC(=CC(=C1)C(=O)OCC=C)C(=O)OCC=C
This item is not returnable. View return policy
17832-16-5
C18H18O6
330.33
MFCD00008647
VOSUIKFOFHZNED-UHFFFAOYSA-N
triallyl trimesate, triallyl benzene-1,3,5-tricarboxylate, triallyl 1,3,5-benzenetricarboxylate, triallylbenzene-1,3,5-tricarboxylate, 1,3,5-benzenetricarboxylic acid, tri-2-propenyl ester, 1,3,5-benzenetricarboxylic acid, 1,3,5-tri-2-propen-1-yl ester, 1,3,5-benzenetricarboxylic acid triallyl ester, 1,3,5-tris prop-2-en-1-yl benzene-1,3,5-tricarboxylate, triallyltrimesate, acmc-1c8hf
87328
tris(prop-2-enyl) benzene-1,3,5-tricarboxylate
C=CCOC(=O)C1=CC(=CC(=C1)C(=O)OCC=C)C(=O)OCC=C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 31°C to 34°C
Color Yellow
Density 1.1600g/mL
Boiling Point 212°C (2.0 mmHg)
Flash Point >113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H3(CO2CH2CH=CH2)3
Quantity 5 g
Specific Gravity 1.16
Formula Weight 330.33
Percent Purity 98%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 1, 3, 5-Benzenetricarboxylic acid triallyl ester
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'