missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Benzenetricarbonyl chloride, 98+%, Thermo Scientific Chemicals

Catalog Number p-7044587
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
4422-95-1
C9H3Cl3O3
265.47
MFCD00000679
UWCPYKQBIPYOLX-UHFFFAOYSA-N
trimesoyl chloride, 1,3,5-benzenetricarbonyl trichloride, 1,3,5-benzenetricarboxylic acid chloride, 1,3,5-benzenetricarbonyl chloride, trimesic acid trichloride, 1,3,5-benzenetricarboxylic chloride, benzene-1,3,5-tricarbonyl trichloride, unii-5866peo79w, 1,5-benzenetricarbonyl trichloride, 1,3,5-benzenetricarboxylicacidchloride
78138
ClC(=O)C1=CC(=CC(=C1)C(Cl)=O)C(Cl)=O
This item is not returnable. View return policy
4422-95-1
C9H3Cl3O3
265.47
MFCD00000679
UWCPYKQBIPYOLX-UHFFFAOYSA-N
trimesoyl chloride, 1,3,5-benzenetricarbonyl trichloride, 1,3,5-benzenetricarboxylic acid chloride, 1,3,5-benzenetricarbonyl chloride, trimesic acid trichloride, 1,3,5-benzenetricarboxylic chloride, benzene-1,3,5-tricarbonyl trichloride, unii-5866peo79w, 1,5-benzenetricarbonyl trichloride, 1,3,5-benzenetricarboxylicacidchloride
78138
ClC(=O)C1=CC(=CC(=C1)C(Cl)=O)C(Cl)=O

1,3,5-Benzenetricarbonyl chloride was used in the synthesis of chiral azoaromatic dendrimeric systems. It was used to study the structure of activated composite membranes containing organophosphorus extractants as carriers. It was the starting material for two tritopic amides derived from 3- and 4-methylaminopyridine which self-assembled into nanoballs on treatment with palladium(II) nitrate in DMSO.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Benzenetricarbonyl chloride was used in the synthesis of chiral azoaromatic dendrimeric systems. It was used to study the structure of activated composite membranes containing organophosphorus extractants as carriers. It was the starting material for two tritopic amides derived from 3- and 4-methylaminopyridine which self-assembled into nanoballs on treatment with palladium(II) nitrate in DMSO.

Solubility
Soluble in chloroform(soluble10%, clear to very slightly hazy, colorless to faintly yellow) and toluene. Solubility in water: Reacts.

Notes
Moisture Sensitive. Store away from strong bases and oxidizing agents. Incompatible with water, bases and oxidizing agents. Store at 4°C.
TRUSTED_SUSTAINABILITY
Melting Point 34°C to 38°C
Boiling Point 180°C (16 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN3261
Beilstein 2940936
Sensitivity Moisture sensitive
Solubility Information Soluble in chloroform(soluble10%,clear to very slightly hazy,colorless to faintly yellow) and toluene. Solubility in water: Reacts.
IUPAC Name benzene-1,3,5-tricarbonyl trichloride
Formula Weight 265.48
Percent Purity ≥98%
Chemical Name or Material 1,3,5-Benzenetricarbonyl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'