missing translation for 'onlineSavingsMsg'
Learn More

1,3,5,7-Cyclooctatetraene, 98%, stabilized, Thermo Scientific Chemicals

Catalog Number 10637934
Click to view available options
Quantity:
250 mg
2.5 g
10 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
629-20-9
C8H8
104.15
MFCD00004161
KDUIUFJBNGTBMD-DLMDZQPMSA-N
1,3,5,7-cyclooctatetraene, 8 annulene, unii-aj19r479cq, cycloocta-1,3,5,7-tetraene, 8-annulene, cyclooctatetraene, 1,5,7-cyclooctatetraene, un2358 flammable liquid, kduiufjbngtbmd-bonzmoemsa-n
637866
cyclooctatetraene
C1=C/C=C\C=C/C=C\1
This item is not returnable. View return policy
629-20-9
C8H8
104.15
MFCD00004161
KDUIUFJBNGTBMD-DLMDZQPMSA-N
1,3,5,7-cyclooctatetraene, 8 annulene, unii-aj19r479cq, cycloocta-1,3,5,7-tetraene, 8-annulene, cyclooctatetraene, 1,5,7-cyclooctatetraene, un2358 flammable liquid, kduiufjbngtbmd-bonzmoemsa-n
637866
cyclooctatetraene
C1=C/C=C\C=C/C=C\1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point -27°C
Color Brown-Yellow to Yellow
Density 0.9200g/mL
Boiling Point 142°C to 143°C
Flash Point 22°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Refractive Index 1.5365 to 1.5385
Quantity 250 mg
Beilstein 05, I, 228
Merck Index 15, 2731
Specific Gravity 0.92
Solubility Information Solubility in water: immiscible
Stabilizer 0.1% hydroquinone
Formula Weight 104.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1,3,5,7-Cyclooctatetraene, Stabilized
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'