Alfa Aesar™ 1-(3,4-Dimethylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea, 97%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H64780.03

UNSPSC: 12352116

Code: NEW

Additional Details:
Additional Details: Weight: 0.00100kg


Quantity 1g

Product Code. 15409146

Quantity Price
1 £ 70.28 / 1g
Estimated Shipment date
from Supplier 14-03-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 1-(3,4-Dimethylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea
Formula Weight 324.3
MDL Number MFCD07058692

Specification

Formula Weight 324.3
Quantity 1g
Chemical Name or Material 1-(3,4-Dimethylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea
MDL Number MFCD07058692
Physical Form Solid
Assay 97%