Alfa Aesar™ 1-(3,4-Dimethylphenyl)-3-(4-fluorophenyl)urea, 97%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H62681.06

5GR 1-(3,4-Dimethylphenyl)-3-(4-fluorophenyl)urea, 97% 5g

UNSPSC: 12352116

Code: NEW

Additional Details:
Additional Details: Weight: 0.00500kg


Quantity 5g

Product Code. 15409178

Quantity Price
1 £ 319.06 / 5g
Estimated Shipment date
from Supplier 06-02-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 1-(3,4-Dimethylphenyl)-3-(4-fluorophenyl)urea
Formula Weight 258.29
MDL Number MFCD06173487

Specification

Formula Weight 258.29
Chemical Name or Material 1-(3,4-Dimethylphenyl)-3-(4-fluorophenyl)urea
Quantity 5g
Physical Form Solid
Assay 97%
MDL Number MFCD06173487