missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(3,4-Difluorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea, 97%

Brand:  Alfa Aesar™ H64349.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15488856

  • £56.92 / 1g


Estimated Shipment date
from Supplier 06-07-2018
Add to basket

Specifications

Chemical Identifiers

1-(3,4-Difluorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea
332.23
MFCD26960699

Specifications

1-(3,4-Difluorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea
Solid
332.23
1g
97%
MFCD26960699
Videos
SDS