missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(3,4-Dichlorophenyl)-3-[4-(trifluoromethyl)phenyl]urea, 97%

Brand:  Alfa Aesar™ H61105.03

Code : NEW

Additional Details : CAS Number : 23788-25-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15416276

  • £73.56 / 1g


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Chemical Identifiers

1-(3,4-Dichlorophenyl)-3-[4-(trifluoromethyl)phenyl]urea
349.14
3068855
23788-25-2
MFCD03416772

Specifications

1-(3,4-Dichlorophenyl)-3-[4-(trifluoromethyl)phenyl]urea
23788-25-2
97%
MFCD03416772
1g
Solid
349.14
3068855
Videos
SDS