missing translation for 'onlineSavingsMsg'
Learn More

1-(3,4-Dichlorobenzyl)piperazine, 97%, Thermo Scientific Chemicals

Catalog Number 15422008
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
55513-17-2
C11H14Cl2N2
245.147
MFCD03407488
PNDSYXGJCWKNFG-UHFFFAOYSA-N
1-3,4-dichlorobenzyl piperazine, 1-3,4-dichlorophenyl methyl piperazine, piperazine, 1-3,4-dichlorophenyl methyl, 1-3,4-dichloro-benzyl-piperazine, acmc-20apc5, 3,4-dichlorobenzylpiperazine, 1-3,4-dichlorobenyl piperazine, 3,4-dichlorophenyl methyl piperazine
2758074
1-[(3,4-dichlorophenyl)methyl]piperazine
C1CN(CCN1)CC2=CC(=C(C=C2)Cl)Cl
This item is not returnable. View return policy
55513-17-2
C11H14Cl2N2
245.147
MFCD03407488
PNDSYXGJCWKNFG-UHFFFAOYSA-N
1-3,4-dichlorobenzyl piperazine, 1-3,4-dichlorophenyl methyl piperazine, piperazine, 1-3,4-dichlorophenyl methyl, 1-3,4-dichloro-benzyl-piperazine, acmc-20apc5, 3,4-dichlorobenzylpiperazine, 1-3,4-dichlorobenyl piperazine, 3,4-dichlorophenyl methyl piperazine
2758074
1-[(3,4-dichlorophenyl)methyl]piperazine
C1CN(CCN1)CC2=CC(=C(C=C2)Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.225 g/mL
Boiling Point 124°C to 128°C (0.02 mmHg)
Flash Point >110°C (230°F)
Quantity 1 g
Formula Weight 245.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-(3,4-Dichlorobenzyl)piperazine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'