missing translation for 'onlineSavingsMsg'
Learn More

1-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)piperidine, 97%, Maybridge

Brand:  Maybridge CC34039DA

Code : 31

Additional Details : CAS Number : 852227-97-5 Weight : 0.05100kg

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10506133

  • £56.00 / 1g


Estimated Shipment date
from Supplier 27-06-2018
Add to basket

Specifications

Chemical Identifiers

1-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperidine
95%
287.21
97%
C17H26BNO2

Specifications

1-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperidine
852227-97-5
97%
C17H26BNO2
1g
Amber Glass Bottle
95%
287.21
Videos
SDS