missing translation for 'onlineSavingsMsg'
Learn More

1-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]piperidine, Thermo Scientific™

Catalog Number p-3176096
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
859833-21-9
C18H28BNO2
301.24
MFCD08435871
KEKNXVTYDJCRMT-UHFFFAOYSA-N
1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl piperidine, 1-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperidine, 1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperidine, piperidine,1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl, 3-piperidine methyl phenylboronic acid pinacol ester, amtb771, 3-piperidin-1-ylmethyl phenylboronic acid, pinacol ester, 2-3-piperidinomethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
16414184
1-[[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]piperidine
CC1(C)OB(OC1(C)C)C1=CC(CN2CCCCC2)=CC=C1
This item is not returnable. View return policy
859833-21-9
C18H28BNO2
301.24
MFCD08435871
KEKNXVTYDJCRMT-UHFFFAOYSA-N
1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl piperidine, 1-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperidine, 1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperidine, piperidine,1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl, 3-piperidine methyl phenylboronic acid pinacol ester, amtb771, 3-piperidin-1-ylmethyl phenylboronic acid, pinacol ester, 2-3-piperidinomethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
16414184
1-[[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]piperidine
CC1(C)OB(OC1(C)C)C1=CC(CN2CCCCC2)=CC=C1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 301.24
Percent Purity 97%
Chemical Name or Material 1-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]piperidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'