missing translation for 'onlineSavingsMsg'
Learn More

1-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]piperidine, Maybridge

Brand:  Maybridge CC45239DA

Code : 31

Additional Details : CAS Number : 859833-21-9 Weight : 0.05100kg Transport : UN number : 2735 Chem class : 8 Pack group : III

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10575164

  • £92.00 / 1g


Estimated Shipment date
from Supplier 28-06-2018
Add to basket

Specifications

Chemical Identifiers

1-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]piperidine
96.5
97%
C18H28BNO2
MFCD08435871
859833-21-9
100.0
97%
301.24

Specifications

1-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]piperidine
859833-21-9
100.0
97%
C18H28BNO2
MFCD08435871
1g
96.5
Amber Glass Bottle
97%
301.24
Videos
SDS