missing translation for 'onlineSavingsMsg'
Learn More

1(2H)-Phthalazinone, 98+%, Thermo Scientific Chemicals

Catalog Number 10483355
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
119-39-1
C8H6N2O
146.15
MFCD00006892
IJAPPYDYQCXOEF-UHFFFAOYSA-N
1 2h-phthalazinone, phthalazin-1 2h-one, phthalazone, phthalazinone, phthalazin-1-one, 1-phthalazinol, phthalazin-1-ol, 1-hydroxyphthalazine, 1 2h phthalazinone, 1,2-dihydrophthalazin-1-one
8394
CHEBI:34023
2H-phthalazin-1-one
C1=CC=C2C(=C1)C=NNC2=O
This item is not returnable. View return policy
119-39-1
C8H6N2O
146.15
MFCD00006892
IJAPPYDYQCXOEF-UHFFFAOYSA-N
1 2h-phthalazinone, phthalazin-1 2h-one, phthalazone, phthalazinone, phthalazin-1-one, 1-phthalazinol, phthalazin-1-ol, 1-hydroxyphthalazine, 1 2h phthalazinone, 1,2-dihydrophthalazin-1-one
8394
CHEBI:34023
2H-phthalazin-1-one
C1=CC=C2C(=C1)C=NNC2=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Melting Point 184.0°C to 187.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Quantity 500 g
Formula Weight 146.15
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material 1(2H)-Phthalazinone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'