missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-[2-(Trifluoromethyl)phenyl]-3-[4-(trifluoromethyl)phenyl]urea, 97%

Brand:  Alfa Aesar™ H61793.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15486776

  • £95.61 / 1g


Estimated Shipment date
from Supplier 07-03-2018
Add to basket

Specifications

Chemical Identifiers

1-[2-(Trifluoromethyl)phenyl]-3-[4-(trifluoromethyl)phenyl]urea
348.24
MFCD06173483

Specifications

1-[2-(Trifluoromethyl)phenyl]-3-[4-(trifluoromethyl)phenyl]urea
1g
97%
MFCD06173483
White
Solid
348.24
Videos
SDS