missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2-Tetrahydrofuroyl)piperazine hydrobromide, 97%

Brand:  Alfa Aesar™ H55490.14

Code : NEW

Additional Details : CAS Number : 63590-62-5 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15492126

  • £212.74 / 25g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

1-(2-Tetrahydrofuroyl)piperazine hydrobromide
63590-62-5
265.15
MFCD03701619

Specifications

1-(2-Tetrahydrofuroyl)piperazine hydrobromide
103° to 107°C
63590-62-5
97%
MFCD03701619
Off-white
25g
Powder
265.15
Videos
SDS