missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Pyridylazo)-2-naphthol, 98%, pure, indicator grade, ACROS Organics™

£63.48 - £346.30

Chemical Identifiers

CAS 85-85-8
Molecular Formula C15H11N3O
Molecular Weight (g/mol) 249.27
InChI Key RAXUMGMWXZYADR-OBGWFSINSA-N
Synonym 1-2-pyridylazo-2-naphthol, unii-095b53y3xv, pan indicator, 2-naphthalenol, 1-2-pyridinylazo, 1e-1-pyridin-2-ylhydrazinylidene naphthalen-2-one, 1-2-pyridylazo-2-hydroxynaphthalene, pan van, 1-2-pyridyldiazenyl naphthalen-2-ol, 1-pyridin-2-azo-2-naphthol, 2-naphthalenol, 1-2-2-pyridinyl diazenyl
PubChem CID 5376264
IUPAC Name (1E)-1-(pyridin-2-ylhydrazinylidene)naphthalen-2-one
SMILES C1=CC=C2C(=C1)C=CC(=O)C2=NNC3=CC=CC=N3
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10637112 SDS View Documents Acros Organics
146310050
5g Glass bottle £63.48
5g View alternative products
Save Up to 
»
In stock, ready to ship: 12 Please to purchase this item. Need a web account? Register with us today!
10657302 SDS View Documents Acros Organics
146310100
10g Glass bottle £108.68
10g View alternative products
Save Up to 
»
In stock, ready to ship: 6 Please to purchase this item. Need a web account? Register with us today!
11996291 SDS View Documents Acros Organics
146310500
50g Glass bottle £346.30
50g View alternative products
Save Up to 
»
Estimated Shipment
28-07-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Indicator
Specifications

Chemical Identifiers

85-85-8
249.27
1-2-pyridylazo-2-naphthol, unii-095b53y3xv, pan indicator, 2-naphthalenol, 1-2-pyridinylazo, 1e-1-pyridin-2-ylhydrazinylidene naphthalen-2-one, 1-2-pyridylazo-2-hydroxynaphthalene, pan van, 1-2-pyridyldiazenyl naphthalen-2-ol, 1-pyridin-2-azo-2-naphthol, 2-naphthalenol, 1-2-2-pyridinyl diazenyl
(1E)-1-(pyridin-2-ylhydrazinylidene)naphthalen-2-one
C15H11N3O
RAXUMGMWXZYADR-OBGWFSINSA-N
5376264
C1=CC=C2C(=C1)C=CC(=O)C2=NNC3=CC=CC=N3

Specifications

1-(2-Pyridylazo)-2-naphthol
Glass bottle
Brown to Orange
5g
98%
22,I,694
1-2-pyridylazo-2-naphthol, unii-095b53y3xv, pan indicator, 2-naphthalenol, 1-2-pyridinylazo, 1e-1-pyridin-2-ylhydrazinylidene naphthalen-2-one, 1-2-pyridylazo-2-hydroxynaphthalene, pan van, 1-2-pyridyldiazenyl naphthalen-2-ol, 1-pyridin-2-azo-2-naphthol, 2-naphthalenol, 1-2-2-pyridinyl diazenyl
RAXUMGMWXZYADR-OBGWFSINSA-N
(1E)-1-(pyridin-2-ylhydrazinylidene)naphthalen-2-one
5376264
Powder
Pure
Authentic
As a complexometric indicator: passes
138.0°C to 141.0°C
85-85-8
C15H11N3O
15,7107
Solubility in water: insoluble.
C1=CC=C2C(=C1)C=CC(=O)C2=NNC3=CC=CC=N3
249.27
249.27
98%