missing translation for 'onlineSavingsMsg'
Learn More

1,2-Phthalic dicarboxaldehyde, 98+%, Thermo Scientific Chemicals

Catalog Number 10755781
39.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass Bottle
643-79-8
C8H6O2
134.13
MFCD00003335
ZWLUXSQADUDCSB-UHFFFAOYSA-N
o-phthalaldehyde, o-phthaldialdehyde, benzene-1,2-dicarboxaldehyde, 1,2-benzenedicarboxaldehyde, phthaldialdehyde, phthalic aldehyde, phthalic dialdehyde, phthalyldicarboxaldehyde, ortho-phthalaldehyde, o-phthaldehyde
4807
CHEBI:70851
phthalaldehyde
O=CC1=CC=CC=C1C=O
643-79-8
C8H6O2
134.13
MFCD00003335
ZWLUXSQADUDCSB-UHFFFAOYSA-N
o-phthalaldehyde, o-phthaldialdehyde, benzene-1,2-dicarboxaldehyde, 1,2-benzenedicarboxaldehyde, phthaldialdehyde, phthalic aldehyde, phthalic dialdehyde, phthalyldicarboxaldehyde, ortho-phthalaldehyde, o-phthaldehyde
4807
CHEBI:70851
phthalaldehyde
O=CC1=CC=CC=C1C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Melting Point 54°C to 57°C
Color White
Boiling Point 83°C to 84°C (1.0 hPa)
Flash Point 132°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass Bottle
Linear Formula C6H4(CHO)2
Quantity 5 g
Beilstein 07, 674
Merck Index 15, 7480
Solubility Information Solubility in water: soluble
Viscosity 2.3 mPa.s (65°C)
Formula Weight 134.13
Percent Purity 98+%
Physical Form Powder
Chemical Name or Material 1, 2-Phthalic dicarboxaldehyde
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'