missing translation for 'onlineSavingsMsg'
Learn More

1,2-Phenylenediacetic Acid 99%, ACROS Organics™

Brand:  Acros Organics 157550250

Code : 29

Additional Details : CAS Number : 7500-53-0 Weight : 0.07500kg


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10582171

  • £101.00 / 25g


EU Stock 2
Add to basket


Specifications

Chemical Identifiers

1, 2-Phenylenediacetic acid
7500-53-0
100.0
99%
C6H4(CH2CO2H)2
MFCD00004329
99%
98.5
≥98.5%
C10H10O4
194.19

Specifications

1, 2-Phenylenediacetic acid
150° to 152°C
99%
98.5
Authentic
Needle-Like Crystalline Powder
99%
C6H4(CH2CO2H)2
MFCD00004329
Beige to beige-brown
25g
7500-53-0
100.0
Glass bottle
≥98.5%
C10H10O4
194.19
09, 874
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 1,2-Phenylenediacetic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS