missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2-Nitrophenyl)pyrrolidine, 97%

Brand:  Alfa Aesar™ H63984.03

Code : NEW

Additional Details : CAS Number : 40832-79-9 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15450979

  • £40.60 / 1g


Estimated Shipment date
from Supplier 30-01-2018
Add to basket

Specifications

Chemical Identifiers

1-(2-Nitrophenyl)pyrrolidine
40832-79-9
192.21
MFCD01604393

Specifications

1-(2-Nitrophenyl)pyrrolidine
Solid
192.21
1g
40832-79-9
97%
MFCD01604393
Videos
SDS