missing translation for 'onlineSavingsMsg'
Learn More

1-[2-(Morpholin-4-yl)ethyl]piperazine, 99%, Thermo Scientific Chemicals

Catalog Number p-2689499
37.20 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
4892-89-1
C10H21N3O
199.29
MFCD00191217
SAJZEJMFAWZNCQ-UHFFFAOYSA-N
4-2-piperazin-1-yl-ethyl-morpholine, 4-2-piperazin-1-yl ethyl morpholine, 4-2-piperazin-1-ylethyl morpholine, 1-2-morpholin-4-yl-ethyl-piperazine, 1-2-morpholinoethyl-piperazine, 1-2-morpholin-4-yl ethyl piperazine, morpholine, 4-2-1-piperazinyl ethyl, 1-2-morpholinoethyl piperazine, 4-2-1-piperazinyl ethyl morpholine
2734640
4-(2-piperazin-1-ylethyl)morpholine
C1CN(CCN1)CCN2CCOCC2
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
4892-89-1
C10H21N3O
199.29
MFCD00191217
SAJZEJMFAWZNCQ-UHFFFAOYSA-N
4-2-piperazin-1-yl-ethyl-morpholine, 4-2-piperazin-1-yl ethyl morpholine, 4-2-piperazin-1-ylethyl morpholine, 1-2-morpholin-4-yl-ethyl-piperazine, 1-2-morpholinoethyl-piperazine, 1-2-morpholin-4-yl ethyl piperazine, morpholine, 4-2-1-piperazinyl ethyl, 1-2-morpholinoethyl piperazine, 4-2-1-piperazinyl ethyl morpholine
2734640
4-(2-piperazin-1-ylethyl)morpholine
C1CN(CCN1)CCN2CCOCC2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color White to Yellow
Boiling Point 82°C to 84°C (0.5 mbar)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 199.29
Percent Purity 99%
Physical Form Oil or Low Melting Crystals
Chemical Name or Material 1-[2-(Morpholin-4-yl)ethyl]piperazine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-¢2-(Morpholin-4-yl)ethyl!piperazine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'