missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2-Methylbenzyl)-3-[4-(trifluoromethyl)phenyl]urea, 97%

Brand:  Alfa Aesar™ H61503.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15456556

  • £63.71 / 1g


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Chemical Identifiers

1-(2-Methylbenzyl)-3-[4-(trifluoromethyl)phenyl]urea
308.3
MFCD26143288

Specifications

1-(2-Methylbenzyl)-3-[4-(trifluoromethyl)phenyl]urea
97%
MFCD26143288
1g
Solid
308.3
White
Videos
SDS