missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Methoxyethyl)piperazine, 98%, Thermo Scientific Chemicals

Catalog Number p-2689365
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
13484-40-7
C7H16N2O
144.21
MFCD00191214
BMEMBBFDTYHTLH-UHFFFAOYSA-N
1-2-methoxyethyl piperazine, 1-2-methoxyethyl-piperazine, 1-2-methoxy-ethyl-piperazine, piperazine, 1-2-methoxyethyl, 1-methoxy-2-piperazinylethane, pubchem14209, 2-methoxyethylpiperazine, n-methoxyethyl piperazine, 2-methoxyethyl piperazine, n-methoxyethylylpiperazine
2734638
1-(2-methoxyethyl)piperazine
COCCN1CCNCC1
This item is not returnable. View return policy
13484-40-7
C7H16N2O
144.21
MFCD00191214
BMEMBBFDTYHTLH-UHFFFAOYSA-N
1-2-methoxyethyl piperazine, 1-2-methoxyethyl-piperazine, 1-2-methoxy-ethyl-piperazine, piperazine, 1-2-methoxyethyl, 1-methoxy-2-piperazinylethane, pubchem14209, 2-methoxyethylpiperazine, n-methoxyethyl piperazine, 2-methoxyethyl piperazine, n-methoxyethylylpiperazine
2734638
1-(2-methoxyethyl)piperazine
COCCN1CCNCC1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9700g/mL
Boiling Point 193°C to 194°C
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4730 to 1.475
Quantity 1 g
Specific Gravity 0.97
Formula Weight 144.21
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-(2-Methoxyethyl)piperazine, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 1-(2-Methoxyethyl)piperazine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'