missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2-Isopropylphenyl)-3-[4-(trifluoromethyl)phenyl]urea, 97%

Brand:  Alfa Aesar™ H61523.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15496566

  • £68.26 / 1g


Estimated Shipment date
from Supplier 07-03-2018
Add to basket

Specifications

Chemical Identifiers

1-(2-Isopropylphenyl)-3-[4-(trifluoromethyl)phenyl]urea
322.32
MFCD26143289

Specifications

1-(2-Isopropylphenyl)-3-[4-(trifluoromethyl)phenyl]urea
1g
97%
MFCD26143289
White
Solid
322.32
Videos
SDS