missing translation for 'onlineSavingsMsg'
Learn More

[1-(2-Furylmethyl)piperid-4-yl]methylamine, 95%, Thermo Scientific™

Catalog Number p-3586550
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
725211-89-2
C11H18N2O
194.28
MFCD09837352
GKCIOGOJFDDOJI-UHFFFAOYSA-N
1-2-furylmethyl piperid-4-yl methylamine, 1-2-furylmethyl piperidin-4-yl methylamine, 1-furan-2-ylmethyl piperidin-4-yl methanamine, 1-1-furan-2-ylmethyl piperidin-4-yl methanamine, 1-2-furylmethyl piperidin-4-yl methyl amine, 1-1-furan-2-yl methyl piperidin-4-yl methanamine
11816398
NCC1CCN(CC2=CC=CO2)CC1
This item is not returnable. View return policy
725211-89-2
C11H18N2O
194.28
MFCD09837352
GKCIOGOJFDDOJI-UHFFFAOYSA-N
1-2-furylmethyl piperid-4-yl methylamine, 1-2-furylmethyl piperidin-4-yl methylamine, 1-furan-2-ylmethyl piperidin-4-yl methanamine, 1-1-furan-2-ylmethyl piperidin-4-yl methanamine, 1-2-furylmethyl piperidin-4-yl methyl amine, 1-1-furan-2-yl methyl piperidin-4-yl methanamine
11816398
NCC1CCN(CC2=CC=CO2)CC1
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name 1-{1-[(furan-2-yl)methyl]piperidin-4-yl}methanamine
Formula Weight 194.28
Percent Purity 95%
Chemical Name or Material [1-(2-Furylmethyl)piperid-4-yl]methylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'