missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Furyl)-1,3-butanedione, 96%, Thermo Scientific Chemicals

Catalog Number 10484125
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
25790-35-6
C8H8O3
152.15
MFCD00041759
GPYKJDYMMUIUFG-UHFFFAOYSA-N
1-2-furyl-1,3-butanedione, 2-furoylacetone, 1-furan-2-yl butane-1,3-dione, 1-2-furanyl-1,3-butanedione, 1,3-butanedione, 1-2-furanyl, 1-2-furyl butane-1,3-dione, 1-2-furyl-1,3-butadione, 1-2-furyl-2,3-butanedione, furoyl acetone, furfuryl-1,3-butanedione
12168
1-(furan-2-yl)butane-1,3-dione
CC(=O)CC(=O)C1=CC=CO1
This item is not returnable. View return policy
25790-35-6
C8H8O3
152.15
MFCD00041759
GPYKJDYMMUIUFG-UHFFFAOYSA-N
1-2-furyl-1,3-butanedione, 2-furoylacetone, 1-furan-2-yl butane-1,3-dione, 1-2-furanyl-1,3-butanedione, 1,3-butanedione, 1-2-furanyl, 1-2-furyl butane-1,3-dione, 1-2-furyl-1,3-butadione, 1-2-furyl-2,3-butanedione, furoyl acetone, furfuryl-1,3-butanedione
12168
1-(furan-2-yl)butane-1,3-dione
CC(=O)CC(=O)C1=CC=CO1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Boiling Point 98.0°C (5.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.5735 to 1.5755
Quantity 25 g
Formula Weight 152.15
Percent Purity 96%
Physical Form Liquid After Melting
Chemical Name or Material 1-(2-Furyl)-1, 3-butanedione
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'